Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COLGALT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303421.1 | NP_001290350.1 | 12 | 2534 | ||
NM_015101.3 | NP_055916.1 | 12 | 3020 | |||
XM_011509332.2 | XP_011507634.1 | 13 | 2826 | |||
GenBank mRNA | AB011156.1 | - | 12 | 2945 | ||
AF288389.1 | - | 12 | 2978 | |||
AK095090.1 | - | 7 | 2255 | |||
AK127287.1 | - | 4 | 2526 |