Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPHP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291593.1 | NP_001278522.1 | 12 | 1862 | ||
NM_001291594.1 | NP_001278523.1 | 11 | 1685 | |||
NM_015102.4 | NP_055917.1 | 15 | 2172 | |||
NR_111987.1 | 15 | 2169 | ||||
XM_006710563.3 | XP_006710626.1 | 15 | 2018 | |||
XM_011541213.1 | XP_011539515.1 | 14 | 1939 | |||
XM_011541214.1 | XP_011539516.1 | 14 | 1942 | |||
XM_011541215.1 | XP_011539517.1 | 13 | 1831 | |||
XM_011541216.2 | XP_011539518.1 | 16 | 2646 | |||
XM_011541217.2 | XP_011539519.1 | 15 | 2510 | |||
XM_011541218.2 | XP_011539520.1 | 16 | 2179 | |||
XM_017000996.1 | XP_016856485.1 | 14 | 1939 | |||
XM_017000997.1 | XP_016856486.1 | 15 | 2441 | |||
XM_017000998.1 | XP_016856487.1 | 14 | 1942 | |||
XM_017000999.1 | XP_016856488.1 | 15 | 1937 | |||
XM_017001000.1 | XP_016856489.1 | 14 | 1770 | |||
XM_017001001.1 | XP_016856490.1 | 9 | 1209 | |||
XM_017001002.1 | XP_016856491.1 | 14 | 1942 | |||
XM_017001003.1 | XP_016856492.1 | 7 | 1000 | |||
GenBank mRNA | AB014573.1 | - | 10 | 1272 | ||
AF537130.1 | - | 15 | 2170 | |||
AK124074.1 | - | 11 | 1638 | |||
AK314326.1 | - | 12 | 1836 | |||
AY118228.1 | - | 15 | 2107 | |||
BC040520.1 | - | 15 | 2170 | 5295140 |