Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK8IP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040439.1 | NP_001035529.1 | 6 | 881 | ||
NM_001318852.1 | NP_001305781.1 | 6 | 921 | |||
NM_015133.4 | NP_055948.2 | 6 | 918 | |||
XM_005255190.2 | XP_005255247.1 | 6 | 921 | |||
XM_006720869.2 | XP_006720932.1 | 6 | 2351 | |||
XM_011522429.2 | XP_011520731.1 | 7 | 945 | |||
XM_011522430.2 | XP_011520732.1 | 7 | 945 | |||
XM_011522431.2 | XP_011520733.1 | 7 | 945 | |||
XM_011522432.2 | XP_011520734.1 | 7 | 945 | |||
XM_011522433.2 | XP_011520735.1 | 2 | 1568 | |||
XM_017023079.1 | XP_016878568.1 | 6 | 921 | |||
XM_017023080.1 | XP_016878569.1 | 6 | 921 | |||
XM_017023081.1 | XP_016878570.1 | 5 | 393 | |||
GenBank mRNA | AB028989.2 | - | 6 | 881 | ||
AK024437.1 | - | 5 | 4412 | |||
BC137123.1 | - | 6 | 833 | 9021120 | ||
BC137124.1 | - | 6 | 833 | 9021121 | ||
BC144486.1 | - | 6 | 836 | 9053013 | ||
BC150266.1 | - | 6 | 881 | 9007222 | ||
CN405915.1 | - | 5 | 355 | |||
CR749696.1 | - | 6 | 2352 |