Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMPCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282944.1 | NP_001269873.1 | 2 | 290 | ||
NM_001282946.1 | NP_001269875.1 | 2 | 290 | |||
NM_015160.2 | NP_055975.1 | 2 | 290 | |||
XM_005266059.4 | XP_005266116.1 | 2 | 205 | |||
XM_017014543.1 | XP_016870032.1 | 1 | 123 | |||
GenBank mRNA | AK296617.1 | - | 2 | 202 | ||
AK299304.1 | - | 2 | 222 | |||
AK307206.1 | - | 2 | 203 | |||
AK308104.1 | - | 2 | 205 | |||
BC022949.1 | - | 2 | 182 | |||
BC033103.1 | - | 2 | 199 | |||
BC111399.1 | - | 2 | 205 | 4609491 | ||
BC132724.1 | - | 2 | 229 | 40146746 | ||
BC136599.1 | - | 2 | 229 | 9020589 | ||
D21064.1 | - | 2 | 203 | |||
D50913.2 | - | 2 | 203 |