Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015169.3 | NP_055984.1 | 1 | 649 | ||
GenBank mRNA | AK024716.1 | - | 1 | 639 | ||
BC001811.2 | - | 1 | 634 | 2824034 | ||
BC013043.2 | - | 1 | 644 | 3603972 | ||
D25218.1 | - | 1 | 649 |