Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136115.2 | NP_001129587.1 | 2 | 363 | ||
NM_015176.3 | NP_055991.1 | 2 | 363 | |||
NR_049764.1 | 2 | 363 | ||||
GenBank mRNA | AB007952.1 | - | 2 | 114 | ||
AK001628.1 | - | 2 | 339 | |||
AK303381.1 | - | 2 | 331 | |||
AK307892.1 | - | 2 | 335 | |||
BC031576.1 | - | 2 | 332 | 5188219 | ||
BP249147.1 | - | 2 | 361 | |||
DB136175.1 | - | 2 | 364 |