Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015190.4 | NP_056005.1 | 3 | 483 | ||
GenBank mRNA | AF327347.1 | - | 3 | 390 | ||
AK074420.1 | - | 3 | 419 | |||
AK094162.1 | - | 3 | 2014 | |||
BC136507.1 | - | 3 | 410 | 9020496 | ||
BX646217.1 | - | 3 | 483 |