Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDS5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100399.1 | NP_001093869.1 | 4 | 924 | ||
NM_001100400.1 | NP_001093870.1 | 4 | 924 | |||
XM_011513672.2 | XP_011511974.1 | 4 | 537 | |||
XM_017007928.1 | XP_016863417.1 | 4 | 499 | |||
GenBank mRNA | AB621803.1 | - | 4 | 938 | ||
AF294791.1 | - | 4 | 495 | |||
AK098331.1 | - | 4 | 508 | |||
AY550968.1 | - | 3 | 421 | |||
BC041361.1 | - | 4 | 928 | |||
BC114218.1 | - | 4 | 620 | 5579832 | ||
BC126225.1 | - | 3 | 334 | 8991941 | ||
CR749425.1 | - | 3 | 293 |