Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BOP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015201.4 | NP_056016.1 | 2 | 343 | ||
GenBank mRNA | AB060694.1 | - | 11 | |||
AK024840.1 | - | 2 | 316 | |||
AK290677.1 | - | 2 | 305 | |||
AK315612.1 | - | 2 | 312 | |||
BC001086.2 | - | 1 | 69 | |||
BC005160.2 | - | 1 | 96 | |||
BC007274.1 | - | 2 | 238 | |||
BC013787.1 | - | 2 | 305 | 3872212 | ||
BC013980.2 | - | 2 | 308 | 3633864 | ||
BC017674.2 | - | 2 | 295 | 3905068 | ||
BE280443.1 | - | 341 | ||||
BQ711345.1 | - | 2 | 336 | |||
BQ961780.1 | - | 331 | ||||
D50914.1 | - | 1 | 83 | |||
DA510177.1 | - | 2 | 344 |