Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POFUT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015227.4 | NP_056042.1 | 2 | 212 | ||
NM_133635.4 | NP_598368.2 | 2 | 212 | |||
NR_004858.1 | 2 | 212 | ||||
XM_006723981.3 | XP_006724044.1 | 2 | 225 | |||
XM_011529504.2 | XP_011527806.1 | 2 | 226 | |||
XM_011529505.2 | XP_011527807.1 | 2 | 227 | |||
XM_011529506.2 | XP_011527808.1 | 2 | 225 | |||
XM_011529507.2 | XP_011527809.1 | 2 | 226 | |||
XM_011529508.2 | XP_011527810.1 | 2 | 227 | |||
XM_011529509.2 | XP_011527811.1 | 2 | 228 | |||
XM_011529510.2 | XP_011527812.1 | 2 | 135 | |||
XM_011529511.2 | XP_011527813.1 | 2 | 228 | |||
XM_011529512.2 | XP_011527814.1 | 2 | 228 | |||
XM_011529513.2 | XP_011527815.1 | 2 | 227 | |||
XM_017028306.1 | XP_016883795.1 | 2 | 225 | |||
XM_017028307.1 | XP_016883796.1 | 2 | 228 | |||
XM_017028308.1 | XP_016883797.1 | 2 | 228 | |||
GenBank mRNA | AB023175.1 | - | 2 | 201 | ||
AJ302079.2 | - | 2 | 201 | |||
AJ302080.2 | - | 2 | 201 | |||
AJ575591.1 | - | 2 | 196 | |||
AK290732.1 | - | 2 | 209 | |||
AK294966.1 | - | 2 | 209 | |||
AK298080.1 | - | 2 | 209 | |||
AL110285.1 | - | 2 | 198 | |||
AY066015.2 | - | 2 | 201 | |||
BC064623.1 | - | 2 | 194 | 5727996 | ||
BI560842.1 | - | 2 | 214 | |||
BI669244.1 | - | 2 | 215 | |||
HG380510.1 | - | 2 | 192 | |||
HG380511.1 | - | 2 | 192 |