Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADCY6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015270.4 | NP_056085.1 | 7 | 1592 | ||
XM_006719210.3 | XP_006719273.1 | 7 | 2550 | |||
XM_017018743.1 | XP_016874232.1 | 6 | 2057 | |||
GenBank mRNA | AB007882.2 | - | 7 | 1533 | ||
AF250226.1 | - | 6 | 2091 | |||
BC064923.1 | - | 8 | 1514 |