Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEDD4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144964.1 | NP_001138436.1 | 2 | 350 | ||
NM_001144965.1 | NP_001138437.1 | 2 | 302 | |||
NM_001144967.2 | NP_001138439.1 | 2 | 396 | |||
NM_001144968.1 | NP_001138440.1 | 2 | 292 | |||
NM_001144969.1 | NP_001138441.1 | 2 | 292 | |||
NM_001243960.1 | NP_001230889.1 | 2 | 396 | |||
NM_015277.5 | NP_056092.2 | 2 | 396 | |||
XM_006722428.3 | XP_006722491.1 | 2 | 412 | |||
XM_017025678.1 | XP_016881167.1 | 2 | 411 | |||
GenBank mRNA | AB071179.1 | - | 2 | 227 | ||
AF385931.1 | - | 2 | 351 | |||
AY112983.1 | - | 2 | 302 | |||
AY112984.2 | - | 2 | 292 | |||
AY112985.2 | - | 2 | 296 | |||
AY256662.1 | - | 3 | 235 | |||
AY312514.1 | - | 2 | 113 | |||
BC032597.1 | - | 2 | 423 | 5528964 | ||
DB473368.1 | - | 2 | 386 |