Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCSTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290184.1 | NP_001277113.1 | 4 | 511 | ||
NM_001290186.1 | NP_001277115.1 | 3 | 372 | |||
NM_015331.2 | NP_056146.1 | 3 | 372 | |||
XM_005245053.4 | XP_005245110.1 | 3 | 589 | |||
XM_011509363.2 | XP_011507665.1 | 3 | 589 | |||
GenBank mRNA | AF240468.1 | - | 3 | 390 | ||
AK299142.1 | - | 3 | 259 | |||
AK310741.1 | - | 4 | 452 | |||
AK314764.1 | - | 3 | 279 | |||
AY359120.1 | - | 3 | 249 | |||
BC047621.1 | - | 4 | 468 | 4827966 | ||
D87442.1 | - | 3 | 246 |