Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPE1-MOB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202485.1 | NP_001189414.1 | 4 | 743 | ||
GenBank mRNA | BJ992310.1 | - | 4 | 361 | ||
CR990871.1 | - | 4 | 327 |
Gene Symbol: MOB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100819.2 | NP_001094289.1 | 2 | 176 | ||
NM_001204094.1 | NP_001191023.1 | 3 | 193 | |||
NM_015387.4 | NP_056202.2 | 3 | 239 | |||
NM_199482.3 | NP_955776.1 | 3 | 249 | |||
GenBank mRNA | AB015441.1 | - | 3 | 198 | ||
AF093825.1 | - | 3 | 214 | |||
AF151853.1 | - | 3 | 214 | |||
AF250319.1 | - | 3 | 224 | |||
AJ580638.1 | - | 3 | 195 | |||
AK027043.1 | - | 3 | 236 | |||
AK289377.1 | - | 3 | 229 | |||
AK292938.1 | - | 3 | 235 | |||
AK297514.1 | - | 2 | 135 | |||
AL080070.1 | - | 3 | 208 | |||
BC005237.1 | - | 3 | 239 | 3996585 | ||
CR457371.1 | - | 3 | 195 | |||
DC298610.1 | - | 3 | 249 | |||
DC350915.1 | - | 2 | 172 | |||
DC375649.1 | - | 3 | 195 |