Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIPF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015388.3 | NP_056203.2 | 2 | 279 | ||
XM_005248989.4 | XP_005249046.1 | 2 | 315 | |||
XM_005248990.2 | XP_005249047.1 | 2 | 320 | |||
XM_011514458.2 | XP_011512760.1 | 2 | 422 | |||
GenBank mRNA | AF162672.1 | - | 2 | 194 | ||
AK000946.1 | - | 2 | 215 | |||
AK021433.1 | - | 191 | ||||
AK021655.1 | - | 2 | 159 | |||
AK022757.1 | - | 2 | 178 | |||
AK298150.1 | - | 2 | 315 | |||
AK308961.1 | - | 2 | 314 | |||
AL050274.1 | - | 2 | 80 | |||
BC019297.1 | - | 2 | 228 | 2905449 | ||
BM836904.1 | - | 2 | 253 | |||
DC387416.1 | - | 2 | 279 |