Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP3A7-CYP3A51P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256497.2 | NP_001243426.2 | 8 | 890 | ||
GenBank mRNA | AF315325.1 | - | 8 | 783 |
Gene Symbol: CYP3A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000765.4 | NP_000756.3 | 8 | 890 | ||
GenBank mRNA | BC067436.1 | - | 8 | 868 | 7002135 | |
BX647143.1 | - | 5 | 1122 | |||
D00408.1 | - | 8 | 786 |