Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145102.1 | NP_001138574.1 | 9 | 4794 | ||
NM_001145103.1 | NP_001138575.1 | 9 | 4605 | |||
NM_001145104.1 | NP_001138576.1 | 7 | 4238 | |||
NM_005902.3 | NP_005893.1 | 9 | 5029 | |||
XM_011521559.2 | XP_011519861.1 | 8 | 5059 | |||
XM_011521560.1 | XP_011519862.1 | 9 | 4971 | |||
GenBank mRNA | AF151027.1 | - | 1 | 277 | ||
AK313812.1 | - | 1 | 525 | |||
AL110265.1 | - | 1 | 832 | |||
BC000414.1 | - | 1 | 182 | |||
BC007496.1 | - | 1 | 182 | |||
CR457386.1 | - | 1 | 106 | |||
CR749287.1 | - | 1 | 3789 |