Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL6IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001313858.1 | NP_001300787.1 | 6 | 710 | ||
NM_015161.2 | NP_055976.1 | 6 | 786 | |||
GenBank mRNA | AK291137.1 | - | 6 | 652 | ||
AK300869.1 | - | 6 | 499 | |||
AK301636.1 | - | 6 | 710 | |||
AK307315.1 | - | 5 | 537 | |||
AK308092.1 | - | 7 | 570 | |||
AK308744.1 | - | 3 | 232 | |||
BC010281.1 | - | 6 | 643 | 3048642 | ||
BX648446.1 | - | 7 | 573 | |||
D31885.1 | - | 6 | 643 |