Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015193.4 | NP_056008.1 | 2, 3 | 2128 | ||
GenBank mRNA | AF193421.1 | - | 2, 3 | 2116 | ||
BC012321.1 | - | 2, 3 | 2132 | 4555433 | ||
BC075802.1 | - | 2, 3 | 2120 | 5198843 | ||
D87468.1 | - | 2, 3 | 2120 |