Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015208.4 | NP_056023.3 | 4 | 519 | ||
XM_005258092.3 | XP_005258149.1 | 4 | 652 | |||
XM_005258093.3 | XP_005258150.1 | 4 | 636 | |||
XM_005258095.2 | XP_005258152.1 | 4 | 491 | |||
XM_011525638.2 | XP_011523940.1 | 4 | 491 | |||
XM_011525639.2 | XP_011523941.1 | 4 | 608 | |||
XM_017025661.1 | XP_016881150.1 | 4 | 620 | |||
XM_017025662.1 | XP_016881151.1 | 4 | 665 | |||
XM_017025667.1 | XP_016881156.1 | 4 | 433 | |||
XM_017025668.1 | XP_016881157.1 | 4 | 433 | |||
GenBank mRNA | AF317425.1 | - | 3 | 255 | ||
AK024808.1 | - | 4 | 469 | |||
AY373757.1 | - | 4 | 469 | |||
BC057225.1 | - | 4 | 468 | |||
BP230422.1 | - | 4 | 519 | |||
BP255733.1 | - | 4 | 512 |