Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPIP5K2 SIRNA Assays ›
Gene Symbol:
PPIP5K2
SLC44A5
Gene Name:
diphosphoinositol pentakisphosphate kinase 2
solute carrier family 44 member 5
Gene Aliases:
CFAP160, HISPPD1, IP7K2, VIP2
CTL5
Chromosome Location:
Chr.5: 103120254 - 103203523 on Build GRCh38
Chromosome Location:
Chr.1: 75202131 - 75724261 on Build GRCh38
UniGene ID:
Hs.212046
Hs.654821
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
22740
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPIP5K2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001276277.1
NP_001263206.1
2, 3
629
NM_001281471.1
NP_001268400.1
2, 3
629
NM_015216.3
NP_056031.2
1, 2
394
XM_005271935.1
XP_005271992.1
2, 3
628
XM_005271936.1
XP_005271993.1
2, 3
628
XM_005271938.1
XP_005271995.1
2, 3
628
XM_005277534.1
XP_005277591.1
2, 3
628
XM_005277535.1
XP_005277592.1
2, 3
628
XM_005277536.1
XP_005277593.1
2, 3
628
XM_005277538.1
XP_005277595.1
2, 3
628
XM_005277539.1
XP_005277596.1
2, 3
628
XM_005277541.1
XP_005277598.1
2, 3
628
XM_006714576.1
XP_006714639.1
2, 3
628
XM_006714577.1
XP_006714640.1
2, 3
628
XM_011543290.2
XP_011541592.1
2, 3
628
XM_017009269.1
XP_016864758.1
2, 3
628
XM_017009270.1
XP_016864759.1
2, 3
628
XM_017009271.1
XP_016864760.1
2, 3
628
XM_017009272.1
XP_016864761.1
2, 3
628
XM_017009273.1
XP_016864762.1
2, 3
628
XM_017009274.1
XP_016864763.1
2, 3
628
XM_017009275.1
XP_016864764.1
2, 3
628
GenBank mRNA
AB007893.1
-
2, 3
619
AK294787.1
-
2, 3
493
BC024591.1
-
1, 2
147
3913521
Gene Symbol: SLC44A5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
GenBank mRNA
CR936677.1
-
4
735
Back To Top
More Information
Panther Classification:
Molecular Function -
nucleotide kinase
Gene Ontology Categories:
Function(s)
Process(es)
inositol metabolic process
phosphorylation
dephosphorylation
inositol phosphate biosynthetic process
inositol phosphate metabolic process
inositol-1,3,4,5,6-pentakisphosphate kinase activity
inositol hexakisphosphate kinase activity
inositol heptakisphosphate kinase activity
inositol hexakisphosphate 5-kinase activity
acid phosphatase activity
ATP binding
diphosphoinositol-pentakisphosphate kinase activity
inositol hexakisphosphate 1-kinase activity
inositol hexakisphosphate 3-kinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline