Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013839.3 | NP_001013861.1 | 3 | 325 | ||
NM_001145297.3 | NP_001138769.1 | 3 | 325 | |||
NM_001145298.3 | NP_001138770.1 | 3 | 325 | |||
NM_001145299.3 | NP_001138771.1 | 3 | 325 | |||
NM_001282313.1 | NP_001269242.1 | 2 | 315 | |||
NM_001282314.1 | NP_001269243.1 | 3 | 325 | |||
NM_015219.4 | NP_056034.2 | 3 | 325 | |||
XM_006721786.3 | XP_006721849.1 | 3 | 324 | |||
XM_006721787.3 | XP_006721850.1 | 3 | 759 | |||
XM_006721788.3 | XP_006721851.1 | 3 | 324 | |||
XM_006721789.3 | XP_006721852.1 | 3 | 324 | |||
XM_011524571.2 | XP_011522873.1 | 1 | 104 | |||
XM_011524572.2 | XP_011522874.1 | 3 | 324 | |||
GenBank mRNA | AB028990.1 | - | 3 | 248 | ||
AK022397.1 | - | 3 | 281 | |||
AK023832.1 | - | 3 | 233 | |||
AK295871.1 | - | 2 | 267 | |||
AL834324.1 | - | 3 | 296 | |||
AY869728.1 | - | 1 | 94 | |||
BC011045.1 | - | 3 | 259 | 4215228 | ||
BC018466.1 | - | 3 | 233 | 3867492 | ||
BX647763.1 | - | 3 | 326 | |||
CB989148.1 | - | 3 | 340 | |||
DA224278.1 | - | 3 | 286 | |||
DC319923.1 | - | 2 | 315 | |||
FJ457119.1 | - | 3 | 230 | |||
FJ457120.1 | - | 3 | 230 | |||
HY020675.1 | - | 3 | 327 |