Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM208A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112736.1 | NP_001106207.1 | 14 | 1631 | ||
NM_015224.3 | NP_056039.2 | 7 | 982 | |||
XM_005264999.1 | XP_005265056.1 | 14 | 1766 | |||
XM_006713077.2 | XP_006713140.1 | 14 | 1766 | |||
XM_006713078.2 | XP_006713141.1 | 8 | 620 | |||
XM_011533552.2 | XP_011531854.1 | 14 | 1766 | |||
XM_011533553.2 | XP_011531855.1 | 13 | 1187 | |||
XM_017006030.1 | XP_016861519.1 | 14 | 1541 | |||
XM_017006031.1 | XP_016861520.1 | 14 | 1766 | |||
XM_017006032.1 | XP_016861521.1 | 13 | 1187 | |||
GenBank mRNA | AF180425.2 | - | 7 | 1266 | ||
AK094486.1 | - | 14 | 1761 | |||
AK307599.1 | - | 6 | 444 | |||
BC029354.1 | - | 7 | 982 | |||
BC070096.1 | - | 14 | 1629 | |||
BC129987.1 | - | 6 | 407 | 40129237 | ||
BX648955.1 | - | 13 | 1113 |