Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040118.2 | NP_001035207.1 | 13 | 1978 | ||
NM_001135190.1 | NP_001128662.1 | 11 | 1789 | |||
NM_015242.4 | NP_056057.2 | 11 | 1789 | |||
GenBank mRNA | AB018325.2 | - | 11 | 1766 | ||
AF411983.1 | - | 7 | 847 | |||
AJ621557.1 | - | 11 | 1789 | |||
AK160383.1 | - | 10 | 2127 | |||
AY049732.1 | - | 8 | 1626 | |||
AY553630.1 | - | 13 | 1993 | |||
BC140792.1 | - | 11 | 1459 | 9021660 | ||
BC144562.1 | - | 11 | 1459 | 9053090 |