Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198665.1 | NP_001185594.1 | 5 | 617 | ||
NM_001301084.1 | NP_001288013.1 | 6 | 500 | |||
NM_015313.2 | NP_056128.1 | 6 | 674 | |||
XM_006718805.3 | XP_006718868.1 | 6 | 1033 | |||
XM_011542720.2 | XP_011541022.1 | 6 | 615 | |||
XM_017017420.1 | XP_016872909.1 | 5 | 1250 | |||
XM_017017421.1 | XP_016872910.1 | 6 | 890 | |||
XM_017017422.1 | XP_016872911.1 | 6 | 502 | |||
GenBank mRNA | AB002380.2 | - | 1 | 36 | ||
AF180681.1 | - | 6 | 346 | |||
AK294803.1 | - | 6 | 475 | |||
AK304316.1 | - | 6 | 898 | |||
BC063117.1 | - | 5 | 620 | |||
BX647785.1 | - | 6 | 476 | |||
BX648515.1 | - | 6 | 481 | |||
DC350360.1 | - | 6 | 500 |