Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164603.1 | NP_001158075.1 | 2 | 557 | ||
NM_015338.5 | NP_056153.2 | 2 | 557 | |||
XM_006723727.3 | XP_006723790.1 | 2 | 839 | |||
XM_006723728.3 | XP_006723791.1 | 2 | 166 | |||
XM_006723730.3 | XP_006723793.1 | 2 | 362 | |||
XM_006723732.2 | XP_006723795.1 | 2 | 338 | |||
XM_011528648.2 | XP_011526950.1 | 3 | 484 | |||
XM_011528652.2 | XP_011526954.1 | 3 | 1499 | |||
XM_017027704.1 | XP_016883193.1 | 2 | 1622 | |||
XM_017027705.1 | XP_016883194.1 | 2 | 635 | |||
XM_017027706.1 | XP_016883195.1 | 2 | 839 | |||
GenBank mRNA | AB084280.1 | - | 2 | 113 | ||
AJ438952.2 | - | 2 | 383 | |||
AK122923.1 | - | 2 | 338 | |||
BC064984.1 | - | 2 | 542 | |||
BC100280.1 | - | 2 | 551 | |||
BC137278.1 | - | 3 | 320 | 9021275 | ||
BC137280.1 | - | 3 | 320 | 9021277 | ||
BQ064719.1 | - | 549 |