Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015340.3 | NP_056155.1 | 3 | 364 | ||
XM_005265006.1 | XP_005265063.1 | 2 | 320 | |||
XM_011533554.2 | XP_011531856.1 | 3 | 383 | |||
XM_017006042.1 | XP_016861531.1 | 3 | 383 | |||
GenBank mRNA | AK308146.1 | - | 2 | 317 | ||
BC025989.2 | - | 3 | 371 | 4822546 | ||
BI463427.1 | - | 3 | 374 | |||
D21851.1 | - | 3 | 364 |