Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEPROTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015344.2 | NP_056159.2 | 4 | 428 | ||
GenBank mRNA | AF063605.1 | - | 4 | 405 | ||
AF161461.1 | - | 4 | 361 | |||
AK075363.1 | - | 4 | 372 | |||
AY277595.1 | - | 4 | 400 | |||
AY358674.1 | - | 4 | 400 | |||
BC000642.2 | - | 4 | 411 | 3344487 | ||
CR541735.1 | - | 3 | 321 | |||
CR541760.1 | - | 3 | 321 | |||
DA258969.1 | - | 4 | 429 |