Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUZ12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015355.3 | NP_056170.2 | 4 | 681 | ||
XM_006721794.3 | XP_006721857.1 | 2 | 123 | |||
XM_017024409.1 | XP_016879898.1 | 4 | 569 | |||
GenBank mRNA | AK290014.1 | - | 4 | 652 | ||
BC015704.1 | - | 4 | 523 | 3908024 | ||
BI668506.1 | - | 4 | 695 | |||
D63881.2 | - | 4 | 638 | |||
DR000680.1 | - | 4 | 677 |
Gene Symbol: SUZ12P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC047718.1 | - | 5 | 711 |