Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253912.1 | NP_001240841.1 | 15 | 2826 | ||
NM_015173.3 | NP_055988.2 | 13 | 2544 | |||
XM_005262646.2 | XP_005262703.1 | 16 | 2865 | |||
XM_005262647.2 | XP_005262704.1 | 15 | 2826 | |||
XM_005262649.2 | XP_005262706.1 | 15 | 2318 | |||
XM_006714001.2 | XP_006714064.1 | 4 | 645 | |||
XM_011513659.2 | XP_011511961.1 | 17 | 2925 | |||
XM_011513660.2 | XP_011511962.1 | 16 | 2886 | |||
XM_011513662.2 | XP_011511964.1 | 16 | 2805 | |||
XM_011513663.2 | XP_011511965.1 | 15 | 2766 | |||
XM_011513664.2 | XP_011511966.1 | 16 | 2763 | |||
XM_011513665.2 | XP_011511967.1 | 15 | 2643 | |||
XM_011513666.2 | XP_011511968.1 | 14 | 2604 | |||
XM_011513670.2 | XP_011511972.1 | 2 | 647 | |||
XM_017007918.1 | XP_016863407.1 | 15 | 2724 | |||
XM_017007919.1 | XP_016863408.1 | 14 | 2706 | |||
XM_017007920.1 | XP_016863409.1 | 14 | 2583 | |||
XM_017007921.1 | XP_016863410.1 | 12 | 1859 | |||
XM_017007922.1 | XP_016863411.1 | 17 | 2925 | |||
GenBank mRNA | AB029031.1 | - | 7 | 971 | ||
AB449881.1 | - | 12 | 2186 | |||
AK027355.1 | - | 5 | 743 | |||
AK295746.1 | - | 15 | 2649 | |||
BC014529.2 | - | 11 | 1382 | |||
BC053648.1 | - | 11 | 1357 | |||
BC126979.1 | - | 3 | 659 |