Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDS5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100399.1 | NP_001093869.1 | 33 | 6041 | ||
GenBank mRNA | AB014548.1 | - | 21 | 4043 | ||
AB621803.1 | - | 33 | 6055 | |||
AF294791.1 | - | 33 | 5610 | |||
AK001828.1 | - | 1 | 868 | |||
BC009650.1 | - | 9 | 2626 | |||
BC114218.1 | - | 33 | 5738 | 5579832 |