Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BOP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015201.4 | NP_056016.1 | 8 | 1089 | ||
GenBank mRNA | AB060694.1 | - | 7 | 757 | ||
AK024840.1 | - | 8 | 1062 | |||
AK290677.1 | - | 8 | 1051 | |||
AK293719.1 | - | 7 | 733 | |||
AK310015.1 | - | 6 | 663 | |||
AK315612.1 | - | 8 | 1058 | |||
BC001086.2 | - | 7 | 815 | |||
BC005160.2 | - | 7 | 842 | |||
BC007274.1 | - | 8 | 984 | |||
BC013787.1 | - | 8 | 1051 | 3872212 | ||
BC013980.2 | - | 8 | 1054 | 3633864 | ||
BC017674.2 | - | 8 | 1041 | 3905068 | ||
BC073813.1 | - | 3 | 348 | |||
BC098446.1 | - | 4 | 485 | |||
D50914.1 | - | 7 | 829 |