Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013839.3 | NP_001013861.1 | 19 | 4728 | ||
NM_001145297.3 | NP_001138769.1 | 20 | 4881 | |||
NM_001145298.3 | NP_001138770.1 | 19 | 4704 | |||
NM_001145299.3 | NP_001138771.1 | 20 | 4797 | |||
NM_001282313.1 | NP_001269242.1 | 19 | 4757 | |||
NM_015219.4 | NP_056034.2 | 18 | 4635 | |||
XM_006721786.3 | XP_006721849.1 | 21 | 4835 | |||
XM_006721787.3 | XP_006721850.1 | 20 | 5201 | |||
XM_006721788.3 | XP_006721851.1 | 20 | 4742 | |||
XM_006721789.3 | XP_006721852.1 | 19 | 4673 | |||
XM_011524571.2 | XP_011522873.1 | 20 | 5112 | |||
XM_017024397.1 | XP_016879886.1 | 16 | 4237 | |||
GenBank mRNA | AB028990.1 | - | 19 | 4651 | ||
AK000718.1 | - | 1 | 818 | |||
AK022552.1 | - | 10 | 3419 | |||
AK023832.1 | - | 18 | 4543 | |||
AK128278.1 | - | 6 | 2875 | |||
AL162005.1 | - | 1 | 2170 | |||
BC015165.1 | - | 1 | 1076 | |||
BX648435.1 | - | 3 | 3284 |