Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POFUT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015227.4 | NP_056042.1 | 5 | 721 | ||
NM_133635.4 | NP_598368.2 | 5 | 721 | |||
NR_004858.1 | 5 | 721 | ||||
XM_006723981.3 | XP_006724044.1 | 6 | 794 | |||
XM_011529504.2 | XP_011527806.1 | 7 | 930 | |||
XM_011529505.2 | XP_011527807.1 | 7 | 931 | |||
XM_011529506.2 | XP_011527808.1 | 6 | 869 | |||
XM_011529507.2 | XP_011527809.1 | 6 | 765 | |||
XM_011529508.2 | XP_011527810.1 | 7 | 931 | |||
XM_011529510.2 | XP_011527812.1 | 7 | 839 | |||
XM_017028306.1 | XP_016883795.1 | 5 | 704 | |||
XM_017028309.1 | XP_016883798.1 | 3 | 127 | |||
GenBank mRNA | AB023175.1 | - | 5 | 710 | ||
AJ302079.2 | - | 5 | 710 | |||
AJ302080.2 | - | 5 | 710 | |||
AJ575591.1 | - | 5 | 705 | |||
AK290732.1 | - | 5 | 718 | |||
AL110285.1 | - | 5 | 707 | |||
AY066015.2 | - | 5 | 710 | |||
BC064623.1 | - | 5 | 703 | 5727996 |