Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015230.3 | NP_056045.2 | 21 | 3850 | ||
XM_017007698.1 | XP_016863187.1 | 21 | 4280 | |||
XM_017007699.1 | XP_016863188.1 | 21 | 3735 | |||
XM_017007700.1 | XP_016863189.1 | 21 | 4280 | |||
XM_017007701.1 | XP_016863190.1 | 21 | 4280 | |||
XM_017007702.1 | XP_016863191.1 | 21 | 4280 | |||
GenBank mRNA | AB011152.2 | - | 20 | 3293 | ||
AF411982.1 | - | 10 | 1289 | |||
AF439781.1 | - | 20 | 3672 | |||
AY049733.1 | - | 21 | 3765 | |||
BC105089.1 | - | 8 | 1064 | |||
BC105091.1 | - | 8 | 1064 | |||
BC150258.1 | - | 20 | 3304 |