Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040118.2 | NP_001035207.1 | 34 | 4476 | ||
NM_001135190.1 | NP_001128662.1 | 30 | 4071 | |||
NM_015242.4 | NP_056057.2 | 32 | 4287 | |||
GenBank mRNA | AB018325.2 | - | 32 | 4264 | ||
AF411983.1 | - | 28 | 3345 | |||
AJ621557.1 | - | 32 | 4287 | |||
AJ706973.1 | - | 2 | 135 | |||
AK160383.1 | - | 30 | 4954 | |||
AY049732.1 | - | 29 | 4124 | |||
AY553630.1 | - | 34 | 4491 | |||
BC008315.1 | - | 15 | 1724 | |||
BC056401.1 | - | 19 | 2097 | |||
BC140792.1 | - | 30 | 3741 | 9021660 | ||
BC144562.1 | - | 31 | 3924 | 9053090 |