Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMXL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174116.1 | NP_001167587.1 | 5 | 668 | ||
NM_001174117.1 | NP_001167588.1 | 5 | 668 | |||
NM_015263.3 | NP_056078.2 | 5 | 668 | |||
XM_005254255.1 | XP_005254312.1 | 5 | 731 | |||
XM_005254256.1 | XP_005254313.1 | 5 | 731 | |||
XM_011521402.2 | XP_011519704.1 | 5 | 731 | |||
XM_017022034.1 | XP_016877523.1 | 5 | 404 | |||
GenBank mRNA | AF389880.1 | - | 5 | 650 | ||
BC140781.1 | - | 5 | 533 | 9021649 | ||
BC144539.1 | - | 5 | 533 | 9053066 |