Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015340.3 | NP_056155.1 | 22 | 2996 | ||
XM_005265006.1 | XP_005265063.1 | 21 | 2952 | |||
GenBank mRNA | AK025465.1 | - | 16 | 2296 | ||
AK026113.1 | - | 9 | 1219 | |||
BC025989.2 | - | 22 | 3003 | 4822546 | ||
D21851.1 | - | 22 | 2996 |