Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other R3HDM1 SIRNA Assays ›
Gene Symbol:
R3HDM1
Gene Name:
R3H domain containing 1
Gene Aliases:
R3HDM
Chromosome Location:
Chr.2: 135531429 - 135725270 on Build GRCh38
UniGene ID:
Hs.412462
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
22884
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: R3HDM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282798.1
NP_001269727.1
26
4425
NM_001282799.1
NP_001269728.1
23
4033
NM_001282800.1
NP_001269729.1
23
4082
NM_015361.3
NP_056176.2
26
4417
XM_005263626.3
XP_005263683.1
26
4223
XM_005263627.3
XP_005263684.1
26
4220
XM_005263628.3
XP_005263685.1
25
4154
XM_005263629.3
XP_005263686.1
25
4151
XM_006712389.2
XP_006712452.1
26
4409
XM_011510870.1
XP_011509172.1
27
4511
XM_011510871.1
XP_011509173.1
27
4527
XM_011510872.2
XP_011509174.1
26
4469
XM_011510873.2
XP_011509175.1
27
4463
XM_011510874.2
XP_011509176.1
25
4379
XM_011510875.2
XP_011509177.1
25
4395
XM_011510876.2
XP_011509178.1
27
4325
XM_011510877.2
XP_011509179.1
27
4277
XM_011510878.2
XP_011509180.1
26
4256
XM_011510879.2
XP_011509181.1
26
4208
XM_011510881.1
XP_011509183.1
26
4366
XM_017003718.1
XP_016859207.1
26
4422
XM_017003719.1
XP_016859208.1
25
4401
XM_017003720.1
XP_016859209.1
26
4374
XM_017003721.1
XP_016859210.1
27
4341
XM_017003722.1
XP_016859211.1
24
4277
XM_017003723.1
XP_016859212.1
24
4293
XM_017003724.1
XP_016859213.1
24
4112
XM_017003725.1
XP_016859214.1
28
4603
XM_017003726.1
XP_016859215.1
28
4673
XM_017003727.1
XP_016859216.1
27
4631
XM_017003728.1
XP_016859217.1
25
4106
XM_017003729.1
XP_016859218.1
26
4448
XM_017003730.1
XP_016859219.1
24
4192
XM_017003731.1
XP_016859220.1
27
4522
XM_017003732.1
XP_016859221.1
27
4497
XM_017003733.1
XP_016859222.1
28
4646
XM_017003734.1
XP_016859223.1
29
4806
XM_017003735.1
XP_016859224.1
26
4404
XM_017003736.1
XP_016859225.1
25
4234
GenBank mRNA
AK026987.1
-
8
1908
AK127793.1
-
23
4053
BC041093.1
-
26
4359
5556247
D21852.1
-
23
3987
Back To Top
More Information
Gene Ontology Categories:
Process(es)
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0