Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195797.1 | NP_001182726.1 | 2, 3 | 359 | ||
NM_015364.4 | NP_056179.3 | 3, 4 | 449 | |||
XM_011517508.2 | XP_011515810.1 | 3, 4 | 382 | |||
XM_017013299.1 | XP_016868788.1 | 3, 4 | 382 | |||
XM_017013300.1 | XP_016868789.1 | 2, 3 | 413 | |||
GenBank mRNA | AB018549.1 | - | 3, 4 | 460 | ||
AB446498.1 | - | 3, 4 | 335 | |||
AF168121.1 | - | 3, 4 | 356 | |||
BC020690.1 | - | 3, 4 | 359 | 4767246 | ||
BG435728.1 | - | 3, 4 | 450 |