Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIPF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015388.3 | NP_056203.2 | 9 | 1295 | ||
XM_005248989.4 | XP_005249046.1 | 9 | 1331 | |||
XM_005248990.2 | XP_005249047.1 | 9 | 1336 | |||
XM_011514458.2 | XP_011512760.1 | 9 | 1438 | |||
GenBank mRNA | AF162672.1 | - | 9 | 1210 | ||
AK000946.1 | - | 9 | 1231 | |||
AK021433.1 | - | 1449 | ||||
AK021655.1 | - | 9 | 1175 | |||
AK022757.1 | - | 9 | 1194 | |||
AL050274.1 | - | 9 | 1096 | |||
BC019297.1 | - | 9 | 1244 | 2905449 | ||
BE562740.1 | - | 356 | ||||
CR533541.1 | - | 8 | 1007 |