Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AASDHPPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015423.2 | NP_056238.2 | 2 | 442 | ||
GenBank mRNA | AF136978.1 | - | 2 | 461 | ||
AF151057.1 | - | 2 | 462 | |||
AF151838.1 | - | 2 | 462 | |||
AF201943.1 | - | 2 | 411 | |||
AK293362.1 | - | 2 | 325 | |||
AK312529.1 | - | 2 | 346 | |||
BC015470.1 | - | 2 | 333 | 3914010 | ||
BC016728.1 | - | 2 | 362 | 4081713 |