Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015431.3 | NP_056246.3 | 2, 3 | 572 | ||
GenBank mRNA | AF327057.1 | - | 2, 3 | 572 | ||
AK096188.1 | - | 2, 3 | 454 | |||
BC074748.2 | - | 1, 2 | 116 | |||
BX644877.1 | - | 2, 3 | 128 | |||
DN998591.1 | - | 2, 3 | 571 |