Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYRIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284423.1 | NP_001271352.1 | 4 | 514 | ||
NM_001284424.1 | NP_001271353.1 | 4 | 703 | |||
NM_001284426.1 | NP_001271355.1 | 2 | 324 | |||
NM_015460.3 | NP_056275.2 | 4 | 703 | |||
NR_104316.1 | 4 | 703 | ||||
XM_011533575.1 | XP_011531877.1 | 4 | 675 | |||
GenBank mRNA | AB083783.1 | - | 3 | 416 | ||
AF396687.1 | - | 4 | 552 | |||
AK125334.1 | - | 2 | 190 | |||
AK126013.1 | - | 4 | 657 | |||
AK294714.1 | - | 2 | 314 | |||
AK310972.1 | - | 4 | 675 | |||
AK312412.1 | - | 4 | 514 | |||
AK316068.1 | - | 2 | 324 | |||
BC092512.1 | - | 4 | 702 | 3093499 | ||
BC109312.1 | - | 4 | 531 | 40036097 |