Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1429 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015496.4 | NP_056311.2 | 9 | 2308 | ||
NM_183009.2 | NP_892121.1 | 9 | 2308 | |||
XM_017013306.1 | XP_016868795.1 | 8 | 2088 | |||
XM_017013307.1 | XP_016868796.1 | 8 | 2131 | |||
XM_017013308.1 | XP_016868797.1 | 7 | 2121 | |||
GenBank mRNA | AB037850.1 | - | 9 | 2232 | ||
AF116724.1 | - | 9 | 2232 | |||
AK022906.1 | - | 1 | 79 | |||
AL832487.1 | - | 9 | 2249 | |||
BC003701.1 | - | 2 | 396 | |||
BC069239.1 | - | 2 | 373 | |||
BC112288.1 | - | 9 | 2320 | 8327756 | ||
BC113380.1 | - | 9 | 2308 | 8322432 |