Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NECAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015509.3 | NP_056324.2 | 6 | 597 | ||
NR_024260.1 | 5 | 515 | ||||
GenBank mRNA | AK074858.1 | - | 6 | 554 | ||
AK074880.1 | - | 6 | 567 | |||
AK074923.1 | - | 6 | 540 | |||
AK075013.1 | - | 6 | 574 | |||
AK094805.1 | - | 1 | 1139 | |||
AK290537.1 | - | 5 | 440 | |||
AK314424.1 | - | 5 | 449 | |||
AL050272.1 | - | 5 | 444 | |||
BC002888.2 | - | 6 | 499 | |||
BC067367.1 | - | 4 | 269 | |||
BC084551.1 | - | 2 | 108 | |||
BC110876.1 | - | 6 | 531 | 5553942 |