Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199570.1 | NP_001186499.1 | 1, 2 | 481 | ||
NM_015526.2 | NP_056341.1 | 2, 3 | 397 | |||
GenBank mRNA | AJ427922.1 | - | 2, 3 | 375 | ||
AK090732.1 | - | 2, 3 | 400 | |||
AK094738.1 | - | 2, 3 | 395 | |||
AK289509.1 | - | 1, 2 | 449 | |||
BC013116.1 | - | 2, 3 | 309 | 4214441 | ||
DA398172.1 | - | 1, 2 | 481 | |||
DA406432.1 | - | 2, 3 | 398 |