Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GORASP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201428.1 | NP_001188357.1 | 3 | 236 | ||
NM_015530.4 | NP_056345.3 | 3 | 991 | |||
XM_006712408.2 | XP_006712471.1 | 3 | 314 | |||
GenBank mRNA | AK023082.1 | - | 3 | 263 | ||
AK023201.1 | - | 3 | 276 | |||
AK027349.1 | - | 3 | 190 | |||
AK125761.1 | - | 3 | 991 | |||
AK225810.1 | - | 3 | 273 | |||
AK293640.1 | - | 3 | 264 | |||
AK296698.1 | - | 4 | 300 | |||
AK298020.1 | - | 3 | 270 | |||
AK315508.1 | - | 3 | 292 | |||
AL117430.1 | - | 3 | 202 | |||
BC007770.2 | - | 3 | 225 | 4298251 | ||
BG034906.1 | - | 3 | 315 | |||
DC399516.1 | - | 3 | 238 |