Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199917.1 | NP_001186846.1 | 3 | 354 | ||
NM_001199919.1 | NP_001186848.1 | 2 | 218 | |||
NM_015599.2 | NP_056414.1 | 2 | 218 | |||
XM_017010936.1 | XP_016866425.1 | 2 | 217 | |||
XM_017010938.1 | XP_016866427.1 | 2 | 206 | |||
GenBank mRNA | AB032081.1 | - | 2 | 130 | ||
AF102265.1 | - | 2 | 134 | |||
AF180371.1 | - | 2 | 141 | |||
AK023432.1 | - | 2 | 163 | |||
AK225363.1 | - | 2 | 156 | |||
AK301867.1 | - | 3 | 332 | |||
AK314512.1 | - | 2 | 151 | |||
BC001258.1 | - | 2 | 177 | 3451677 | ||
DB004046.1 | - | 2 | 219 | |||
DB033430.1 | - | 2 | 217 |