Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHCTF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323342.1 | NP_001310271.1 | 15, 16 | 2361 | ||
NM_001323343.1 | NP_001310272.1 | 15, 16 | 2361 | |||
NM_015446.4 | NP_056261.4 | 15, 16 | 2077 | |||
NR_136586.1 | 15, 16 | 2361 | ||||
XM_006711758.1 | XP_006711821.1 | 15, 16 | 2190 | |||
XM_011544156.2 | XP_011542458.2 | 15, 16 | 2056 | |||
XM_011544157.2 | XP_011542459.2 | 15, 16 | 2056 | |||
GenBank mRNA | AB059277.1 | - | 15, 16 | 2077 | ||
AL832213.1 | - | 5, 6 | 489 | |||
AY157619.1 | - | 15, 16 | 2195 |
Gene Symbol: AHCTF1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_077058.1 | 1 | 2177 |